5-(Pyridin-2-yl)-1,2-dihydropyridin-2-one 98%

Supplier: Ambeed
Warning

Synonyms: [2,3'-Bipyridin]-6'(1'H)-one

A232828-5G A232828-1G
76826-518EA 88.06 USD
76826-518 77612-274
5-(Pyridin-2-yl)-1,2-dihydropyridin-2-one 98%
5-(Pyridin-2-yl)-1,2-dihydropyridin-2-one

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O
MW: 172.183 g/mol
MDL Number: MFCD11113414
CAS Number: 381233-78-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR