3-(2-Chloropyrimidin-4-yl)-1H-indole 96%

Supplier: Ambeed
Warning

A232799-1G A232799-5G A232799-250MG
76826-514EA 79.57 USD
76826-514 76826-516 77683-681
3-(2-Chloropyrimidin-4-yl)-1H-indole 96%
3-(2-Chloropyrimidin-4-yl)-1H-indole

Precaución: For research use only.

Formula: C₁₂H₈ClN₃
MW: 229.67 g/mol
MDL Number: MFCD16658942
CAS Number: 945016-63-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR