Methyl-3-amino-4-nitrobenzoate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-4-nitrobenzoate

A232470-100MG A232470-1G A232470-250MG A232470-5G
76826-504EA 14.86 USD
76826-504 76826-506 76826-508 77683-677
Methyl-3-amino-4-nitrobenzoate 97%
Methyl-3-amino-4-nitrobenzoate

Precaución: For research use only.

Formula: C₈H₈N₂O₄
MW: 196.16 g/mol
MDL Number: MFCD00017008
CAS Number: 99512-09-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR