2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine 98%

Supplier: Ambeed
Warning

Synonyms: 2,5-Dichloro-N-(2-(isopropylsulphonyl)phenyl)pyrimidin-4-amine

A232353-10G A232353-1G A232353-250MG A232353-25G A232353-5G
76826-486EA 126.25 USD
76826-486 76826-488 76826-490 76826-492 76826-494
2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine 98%
2,5-Dichloro-N-[2-(isopropylsulfonyl)phenyl]pyrimidin-4-amine

Precaución: for Research Use Only

Formula: C₁₃H₁₃Cl₂N₃O₂S
MW: 346.23 g/mol
MDL Number: MFCD12827956
CAS Number: 761440-16-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR