2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-Isopropyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A231920-100MG A231920-1G A231920-250MG
76826-474EA 65.78 USD
76826-474 76826-476 76826-478
2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester 98%
2-(Isopropylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₂₂BN₃O₂
MW: 263.1400 g/mol
MDL Number: MFCD12546556
CAS Number: 1218791-46-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR