3-Chloro-N-((S)-1-(2-(dimethylamino)acetamido)-3-(4-(8-((S)-1-hydroxyethyl)imidazo[1,2-a]pyridin-2-yl)phenyl)propan-2-yl)-4-isopropoxybenzamide ≥98%

Supplier: Ambeed
Warning

Synonyms: GSK-923295

A231503-10MG A231503-1MG A231503-5MG
76826-458EA 164 USD
76826-458 76826-460 76826-462
3-Chloro-N-((S)-1-(2-(dimethylamino)acetamido)-3-(4-(8-((S)-1-hydroxyethyl)imidazo[1,2-a]pyridin-2-yl)phenyl)propan-2-yl)-4-isopropoxybenzamide ≥98%
3-Chloro-N-((S)-1-(2-(dimethylamino)acetamido)-3-(4-(8-((S)-1-hydroxyethyl)imidazo[1,2-a]pyridin-2-yl)phenyl)propan-2-yl)-4-isopropoxybenzamide

Precaución: for Research Use Only

Formula: C₃₂H₃₈ClN₅O₄
MW: 592.13 g/mol
MDL Number: MFCD16038931
CAS Number: 1088965-37-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR