(S)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binaphthalene]-2,2'-diol
A230826-100MG
A230826-1G
A230826-250MG
A230826-5G
76826-442EA
29
USD
76826-442
76826-444
76826-446
77885-582
(S)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol 98%
(S)-3,3'-Bis(2,4,6-triisopropylphenyl)-[1,1'-binapthalene]-2,2'-diol
Precaución: for Research Use Only
Formula:
C₅₀H₅₈O₂ |
MDL Number:
MFCD11045378 CAS Number: 908338-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...