4-Aminopyrimidin-5-ol 95%

Supplier: Ambeed
Warning

A230172-100MG A230172-1G A230172-5G A230172-250MG
76826-430EA 39 USD
76826-430 76826-432 76826-434 77885-580
4-Aminopyrimidin-5-ol 95%
4-Aminopyrimidin-5-ol

Precaución: for Research Use Only

Formula: C₄H₅N₃O
MDL Number: MFCD18642504
CAS Number: 52601-89-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR