L-allo-Threonine 97%

Supplier: Ambeed
Warning

Synonyms: H-allo-Thr-OH, L(+)-allo-Threonine, Allo-Thr-OH

A229838-100G A229838-10G A229838-1G A229838-250MG A229838-25G A229838-500G A229838-5G
76826-416EA 497.57 USD
76826-416 76826-418 76826-420 76826-422 76826-424 76826-426 76826-428
L-allo-Threonine 97%
L-allo-Threonine

Precaución: for Research Use Only

Formula: C₄H₉NO₃
MW: 119.12 g/mol
MDL Number: MFCD00064268
CAS Number: 28954-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR