3-Amino-6-cyano-2-methylpyridine 98%

Supplier: Ambeed
Warning

A229555-1G A229555-5G
76826-412EA 42 USD
76826-412 76826-414
3-Amino-6-cyano-2-methylpyridine 98%
3-Amino-6-cyano-2-methylpyridine

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD08062828
CAS Number: 1079054-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR