2-(4-Methoxybenzylamino)-5-pyrimidineboronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-Methoxybenzyl)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrimidin-2-amine

A229535-100MG A229535-1G A229535-250MG A229535-5G
76826-404EA 36 USD
76826-404 76826-406 76826-408 76826-410
2-(4-Methoxybenzylamino)-5-pyrimidineboronic acid pinacol ester 98%
2-(4-Methoxybenzylamino)-5-pyrimidineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₈H₂₄BN₃O₃
MW: 341.2100 g/mol
MDL Number: MFCD15143596
CAS Number: 1218790-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR