Diethyl cyclohexane-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A229242-1G A229242-250MG A229242-5G
76826-398EA 11 USD
76826-398 77885-576 77885-577
Diethyl cyclohexane-1,4-dicarboxylate 95%
Diethyl cyclohexane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₂₀O₄
MW: 228.28 g/mol
MDL Number: MFCD12973977
CAS Number: 72903-27-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR