4-Isobutoxyphenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (4-Isobutoxyphenyl)boronic acid

A229082-100G A229082-1G A229082-5G
76826-392EA 513.48 USD
76826-392 76826-394 76826-396
4-Isobutoxyphenylboronic acid 95%
4-Isobutoxyphenylboronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₅BO₃
MW: 194.0400 g/mol
MDL Number: MFCD06798083
CAS Number: 153624-44-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR