1-(4,5-Dihydrothiazol-2-yl)azetidine-3-thiol hydrochloride 95%

Supplier: Ambeed
Warning

A227871-100MG A227871-1G A227871-5G
76826-368EA 6.37 USD
76826-368 76826-370 76826-372
1-(4,5-Dihydrothiazol-2-yl)azetidine-3-thiol hydrochloride 95%
1-(4,5-Dihydrothiazol-2-yl)azetidine-3-thiol hydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁ClN₂S₂
MW: 210.74 g/mol
MDL Number: MFCD18072489
CAS Number: 179337-57-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR