3-(N-(4-Chloro-3-methylisoxazol-5-yl)sulfamoyl)thiophene-2-carboxylic acid 97%

Supplier: Ambeed
Warning

A227692-100MG A227692-1G A227692-250MG
76826-362EA 93 USD
76826-362 76826-364 76826-366
3-(N-(4-Chloro-3-methylisoxazol-5-yl)sulfamoyl)thiophene-2-carboxylic acid 97%
3-(N-(4-Chloro-3-methylisoxazol-5-yl)sulfamoyl)thiophene-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₇ClN₂O₅S₂
MDL Number: MFCD18642943
CAS Number: 184040-74-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR