(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine 99+%

Supplier: Ambeed
Warning

A226184-100MG A226184-1G A226184-250MG A226184-5G
76826-336EA 9 USD
76826-336 76826-338 76826-340 76826-342
(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine 99+%
(1R,2R)-N1,N1-Dimethylcyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₈H₁₈N₂
MW: 142.24 g/mol
MDL Number: MFCD09054929
CAS Number: 320778-92-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR