Pyrazolo[1,5-a]pyridine-5-carboxylic acid 96%

Supplier: Ambeed
Warning

A226169-1G A226169-250MG A226169-5G
76826-330EA 31 USD
76826-330 76826-332 76826-334
Pyrazolo[1,5-a]pyridine-5-carboxylic acid 96%
Pyrazolo[1,5-a]pyridine-5-carboxylic acid

Precaución: for Research Use Only

Formula: C₈H₆N₂O₂
MDL Number: MFCD12828626
CAS Number: 104468-87-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR