1-(2,2,2-Trifluoroethyl)piperazine dihydrochloride 95%

Supplier: Ambeed
Warning

A226165-100MG A226165-1G A226165-250MG A226165-25G A226165-5G
76826-320EA 8 USD
76826-320 76826-322 76826-324 76826-326 76826-328
1-(2,2,2-Trifluoroethyl)piperazine dihydrochloride 95%
1-(2,2,2-Trifluoroethyl)piperazine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₁F₃N₂·2ClH
MDL Number: MFCD08261052
CAS Number: 13349-91-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR