Oxindole-7-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 7-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)indolin-2-one

A225675-1G A225675-250MG A225675-5G
76826-290EA 207.94 USD
76826-290 76826-292 76826-294
Oxindole-7-boronic acid pinacol ester 98%
Oxindole-7-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₃
MW: 259.11 g/mol
MDL Number: MFCD11855966
CAS Number: 1150271-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR