3-Cyano-4-hydroxybenzeneboronic acid pinacol ester 95%

Supplier: Ambeed
Warning

A224236-1G A224236-5G A224236-250MG A224236-25G
76826-264EA 36 USD
76826-264 76826-266 77612-178 77885-559
3-Cyano-4-hydroxybenzeneboronic acid pinacol ester 95%
3-Cyano-4-hydroxybenzeneboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₆BNO₃
MDL Number: MFCD16994355
CAS Number: 775351-56-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR