Methyl 2-(4-(4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butanoyl)phenyl)-2-methylpropanoate 95%
Supplier: Ambeed
|
Warning
|
A224165-1G
A224165-25G
A224165-5G
76826-258EA
15
USD
76826-258
76826-260
76826-262
Methyl 2-(4-(4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butanoyl)phenyl)-2-methylpropanoate 95%
Methyl 2-(4-(4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butanoyl)phenyl)-2-methylpropanoate
Precaución: for Research Use Only
Formula:
C₃₃H₃₉NO₄ |
MDL Number:
MFCD08063765 CAS Number: 154477-55-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...