5-Iodothiophene-2-carbonitrile 95%

Supplier: Ambeed
Warning

A224062-100MG A224062-1G
76826-250EA 97.61 USD
76826-250 76826-252
5-Iodothiophene-2-carbonitrile 95%
5-Iodothiophene-2-carbonitrile

Precaución: for Research Use Only

Formula: C₅H₂INS
MW: 235.05 g/mol
MDL Number: MFCD14584473
CAS Number: 18945-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR