Methyl-5-nitrobenzo[b]thiophene-2-carboxylate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: Methyl 5-nitrobenzo[b]thiophene-2-carboxylate

A223597-100G A223597-10G A223597-1G A223597-250MG A223597-5G
76826-238EA 1267 USD
76826-238 76826-240 76826-242 76826-244 77683-649
Methyl-5-nitrobenzo[b]thiophene-2-carboxylate 95%, Ambeed.Inc
Methyl-5-nitrobenzo[b]thiophene-2-carboxylate

Precaución: For research use only.

Formula: C₁₀H₇NO₄S
MW: 237.2300 g/mol
MDL Number: MFCD03618409
CAS Number: 20699-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR