3,5-Dibromo-4-cyanopyridine 95%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dibromoisonicotinonitrile

A221907-1G A221907-250MG A221907-25G A221907-5G
76826-210EA 15 USD
76826-210 76826-212 76826-214 76826-216
3,5-Dibromo-4-cyanopyridine 95%
3,5-Dibromo-4-cyanopyridine

Precaución: for Research Use Only

Formula: C₆H₂Br₂N₂
MDL Number: MFCD09263321
CAS Number: 870244-34-1
UN: 2923
ADR: 8,6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR