Ethyl-2-methoxyisonicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Ethyl 2-methoxyisonicotinate

A221872-1G A221872-5G
76826-206EA 39.26 USD
76826-206 76826-208
Ethyl-2-methoxyisonicotinate 97%
Ethyl 2-methoxyisonicotinate

Precaución: for Research Use Only

Formula: C₉H₁₁NO₃
MW: 181.1900 g/mol
MDL Number: MFCD11504865
CAS Number: 105596-61-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR