(3-(Benzyloxy)-4-chlorophenyl)boronic acid 98%

Supplier: Ambeed
Warning

A221575-1G A221575-250MG
76826-202EA 64.72 USD
76826-202 76826-204
(3-(Benzyloxy)-4-chlorophenyl)boronic acid 98%
(3-(Benzyloxy)-4-chlorophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₂BClO₃
MW: 262.49 g/mol
MDL Number: MFCD08701723
CAS Number: 1007170-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR