(E/Z)-Locostatin 95%, Ambeed.Inc

Supplier: Ambeed
Warning

Synonyms: (S)-4-Benzyl-3-(but-2-enoyl)oxazolidin-2-one, (E/Z)-UIC-1005

A221051-1G A221051-250MG A221051-100MG A221051-5G
76826-198EA 159.14 USD
76826-198 76826-200 77683-637 77683-638
(E/Z)-Locostatin 95%, Ambeed.Inc
(E/Z)-Locostatin

Precaución: For research use only.

Formula: C₁₄H₁₅NO₃
MW: 245.27 g/mol
MDL Number: MFCD27937442
CAS Number: 133812-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR