6-Bromo-1-(phenylsulfonyl)-1H-indole 98%

Supplier: Ambeed
Warning

A221041-10G A221041-1G A221041-250MG A221041-5G A221041-25G
76826-190EA 78 USD
76826-190 76826-192 76826-194 76826-196 77885-546
6-Bromo-1-(phenylsulfonyl)-1H-indole 98%
6-Bromo-1-(phenylsulfonyl)-1H-indole

Precaución: for Research Use Only

Formula: C₁₄H₁₀BrNO₂S
MDL Number: MFCD03840610
CAS Number: 679794-03-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR