3,5-Dichloropyridine-2-carboxamide 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dichloropicolinamide

A220787-100MG A220787-1G A220787-250MG
76826-184EA 33.95 USD
76826-184 76826-186 76826-188
3,5-Dichloropyridine-2-carboxamide 98%
3,5-Dichloropyridine-2-carboxamide

Precaución: for Research Use Only

Formula: C₆H₄Cl₂N₂O
MW: 191.011 g/mol
MDL Number: MFCD20529142
CAS Number: 5468-71-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR