4-Trimethylsilylphenylboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: Trimethyl(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)silane

A220184-1G A220184-250MG A220184-5G
76826-164EA 46.68 USD
76826-164 76826-166 76826-168
4-Trimethylsilylphenylboronic acid, pinacol ester 98%
4-Trimethylsilylphenylboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₅BO₂Si
MW: 276.254 g/mol
MDL Number: MFCD09037796
CAS Number: 1186026-67-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR