4-Bromo-3-methylbenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A219961-100MG A219961-1G A219961-250MG
76826-144EA 54.11 USD
76826-144 76826-146 76826-148
4-Bromo-3-methylbenzene-1,2-diamine 97%
4-Bromo-3-methylbenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₇H₉BrN₂
MDL Number: MFCD18642364
CAS Number: 952511-74-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR