2,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine ≥95%, ee 99%
Supplier: Ambeed
|
Warning
|
A219077-100MG
A219077-250MG
A219077-1G
76826-122EA
7.43
USD
76826-122
76826-124
77550-424
2,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine ≥95%, ee 99%
2,6-Bis((R)-4-benzyl-4,5-dihydrooxazol-2-yl)pyridine
Precaución: for Research Use Only
Formula:
C₂₅H₂₃N₃O₂ MW: 397.47 g/mol |
MDL Number:
MFCD23704922 CAS Number: 365215-38-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...