(4-Chlorophenyl)(piperazin-1-yl)methanone 95%

Supplier: Ambeed
Warning

A218898-100MG A218898-250MG
76826-118EA 30.77 USD
76826-118 76826-120
(4-Chlorophenyl)(piperazin-1-yl)methanone 95%
(4-Chlorophenyl)(piperazin-1-yl)methanone

Precaución: for Research Use Only

Formula: C₁₁H₁₃ClN₂O
MW: 224.68 g/mol
MDL Number: MFCD01050442
CAS Number: 54042-47-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR