2,4-Dichloro-6-methylnicotinonitrile 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A218813-100MG A218813-1G A218813-250MG A218813-5G
76826-112EA 20 USD
76826-112 76826-114 76826-116 77683-631
2,4-Dichloro-6-methylnicotinonitrile 98%, Ambeed.Inc
2,4-Dichloro-6-methylnicotinonitrile

Precaución: For research use only.

Formula: C₇H₄Cl₂N₂
MW: 187.03 g/mol
MDL Number: MFCD11847537
CAS Number: 38367-36-1
New Product

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR