3,5-Dihydroxybenzoic acid 98%

Supplier: Ambeed
Warning

A218796-1000G A218796-100G A218796-500G A218796-5G A218796-25G
76826-102EA 124 USD
76826-102 76826-104 76826-106 76826-108 76826-110 77088-154
3,5-Dihydroxybenzoic acid 98%
3,5-Dihydroxybenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆O₄
MDL Number: MFCD00002512
CAS Number: 99-10-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR