3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid 96%

Supplier: Ambeed
Warning

Synonyms: (3-((2-(Piperidin-1-yl)ethyl)carbamoyl)phenyl)boronic acid

A218667-100MG A218667-1G A218667-250MG A218667-5G
76826-090EA 39 USD
76826-090 76826-092 76826-094 76826-096
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid 96%
3-(2-(Piperidin-1-yl)ethylcarbamoyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₄H₂₁BN₂O₃
MDL Number: MFCD11053842
CAS Number: 1072946-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR