Methyl 4-iodo-1H-pyrazole-3-carboxylate 98%

Supplier: Ambeed
Warning

A217386-100MG A217386-1G A217386-250MG A217386-5G
76826-078EA 17 USD
76826-078 76826-080 76826-082 77885-541
Methyl 4-iodo-1H-pyrazole-3-carboxylate 98%
Methyl 4-iodo-1H-pyrazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₅IN₂O₂
MDL Number: MFCD09473436
CAS Number: 136944-79-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR