4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane 99%
Supplier: Ambeed
|
Warning
|
A216488-100G
A216488-10G
A216488-1G
A216488-25G
A216488-5G
76826-052EA
129
USD
76826-052
76826-054
76826-056
76826-058
76826-060
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane 99%
4,4-Dimethyl-3,5,8-trioxabicyclo[5.1.0]octane
Precaución: for Research Use Only
Formula:
C₇H₁₂O₃ MW: 144.17 g/mol |
MDL Number:
MFCD16621162 CAS Number: 57280-22-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...