Thiomorpholine-1,1-dioxide 95%

Supplier: Ambeed
Warning

A216210-100G A216210-10G A216210-1G A216210-25G A216210-5G
76826-036EA 486 USD
76826-036 76826-038 76826-040 76826-042 76826-044 76826-046
Thiomorpholine-1,1-dioxide 95%
Thiomorpholine 1,1-dioxide

Precaución: for Research Use Only

Formula: C₄H₉NO₂S
MDL Number: MFCD05861623
CAS Number: 39093-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR