(R)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol ≥98%, ee 99%

Supplier: Ambeed
Warning

Synonyms: (R)-3,3'-Bis(3,5-dimethylphenyl)-[1,1'-binaphthalene]-2,2'-diol

A216163-100MG A216163-1G A216163-250MG A216163-5G
76826-028EA 92.3 USD
76826-028 76826-030 76826-032 76826-034
(R)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol ≥98%, ee 99%
(R)-3,3'-Bis(3,5-dimethylphenyl)-1,1'-bi-2-naphthalene]-2,2'-diol

Precaución: for Research Use Only

Formula: C₃₆H₃₀O₂
MW: 494.622 g/mol
MDL Number: MFCD11617327
CAS Number: 215433-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR