Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate 97%

Supplier: Ambeed
Warning

A215624-25G A215624-5G A215624-10G
76826-024EA 70 USD
76826-024 76826-026 77885-533
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate 97%
Ethyl-3-carbamimidoylamino-4-methylbenzoate nitrate

Precaución: for Research Use Only

Formula: C₁₁H₁₅N₃O₂
MDL Number: MFCD11100711
CAS Number: 641569-96-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR