4,5-Dichloro-2-methylpyrimidine 96%

Supplier: Ambeed
Warning

A215272-1G A215272-250MG A215272-10G
76826-020EA 37 USD
76826-020 76826-022 77885-532
4,5-Dichloro-2-methylpyrimidine 96%
4,5-Dichloro-2-methylpyrimidine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂
MDL Number: MFCD16987924
CAS Number: 26740-71-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR