Methyl 6-oxopiperidine-2-carboxylate 97%

Supplier: Ambeed
Warning

A215081-100MG A215081-1G A215081-250MG A215081-5G
76826-006EA 74.27 USD
76826-006 76826-008 76826-010 76826-012
Methyl 6-oxopiperidine-2-carboxylate 97%
Methyl 6-oxopiperidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.16 g/mol
MDL Number: MFCD11111644
CAS Number: 111479-60-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR