1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%

Supplier: Ambeed
Warning

A215075-100MG A215075-1G A215075-250MG A215075-5G
76825-998EA 12.74 USD
76825-998 76826-000 76826-002 76826-004
1-(2-Chlorophenyl)cyclopropanecarboxylic acid 97%
1-(2-Chlorophenyl)cyclopropanecarboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₉ClO₂
MW: 196.63 g/mol
MDL Number: MFCD07374435
CAS Number: 122143-19-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR