(2,6-Dibromophenyl)methanol 98%

Supplier: Ambeed
Warning

A214973-100MG A214973-10G A214973-1G A214973-250MG A214973-25G A214973-5G
76825-986EA 5 USD
76825-986 76825-988 76825-990 76825-992 76825-994 76825-996
(2,6-Dibromophenyl)methanol 98%
(2,6-Dibromophenyl)methanol

Precaución: for Research Use Only

Formula: C₇H₆Br₂O
MDL Number: MFCD13185492
CAS Number: 1013031-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR