1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid 96%
Supplier: Ambeed
|
Warning
|
A214374-100MG
A214374-1G
A214374-5G
76825-970EA
40
USD
76825-970
76825-972
76825-974
1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid 96%
1-Methyl-5-phenyl-1H-pyrazole-3-carboxylic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₀N₂O₂ |
MDL Number:
MFCD08271933 CAS Number: 10199-53-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...