2-Amino-4-chloro-5-sulfamoylbenzoic acid 95%

Supplier: Ambeed
Warning

A214008-50MG A214008-1G A214008-250MG
76825-966EA 76 USD
76825-966 77885-524 77885-525
2-Amino-4-chloro-5-sulfamoylbenzoic acid 95%
2-Amino-4-chloro-5-sulfamoylbenzoic acid

Precaución: for Research Use Only

Formula: C₇H₇ClN₂O₄S
MDL Number: MFCD03423689
CAS Number: 3086-91-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR