4-(Isopropylamino)benzeneboronic acid, pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: N-Isopropyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline, N-(Prop-2-yl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)aniline

A212905-1G A212905-250MG A212905-5G
76825-942EA 28 USD
76825-942 76825-944 76825-946
4-(Isopropylamino)benzeneboronic acid, pinacol ester 97%
4-(Isopropylamino)benzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₅H₂₄BNO₂
MDL Number: MFCD17015838
CAS Number: 1256360-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR