tert-Butyl-3-ethylpyridin-2-ylcarbamate ≥95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl (3-ethylpyridin-2-yl)carbamate

A212624-100MG A212624-1G A212624-250MG
76825-936EA 14.86 USD
76825-936 76825-938 76825-940
tert-Butyl-3-ethylpyridin-2-ylcarbamate ≥95%
tert-Butyl-3-ethylpyridin-2-ylcarbamate

Precaución: for Research Use Only

Formula: C₁₂H₁₈N₂O₂
MW: 222.29 g/mol
MDL Number: MFCD11976363
CAS Number: 149489-03-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR