3-Fluoro-5-nitropyridine 97%

Supplier: Ambeed
Warning

A211680-10G A211680-1G A211680-5G A211680-250MG
76825-924EA 676.86 USD
76825-924 76825-926 76825-928 77593-388
3-Fluoro-5-nitropyridine 97%
3-Fluoro-5-nitropyridine

Precaución: for Research Use Only

Formula: C₅H₃FN₂O₂
MW: 142.088 g/mol
MDL Number: MFCD13189114
CAS Number: 1060804-39-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR